Product Details

SNP ID
rs146038371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:22769282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATGTCCAGGCATTGCCACTCCT[C/T]CAGAGCGAATTCTATGGTCACATCC
Phenotype
MIM: 603981
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF99 PubMed Links

Gene Details

Gene
ZNF99
Gene Name
zinc finger protein 99
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080409.2 Intron NP_001073878.2

View Full Product Details