Product Details
- SNP ID
-
rs146081922
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57590252 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAGTGCAAAGAAATCCTTGAAGAG[A/G]GACATGGACAGAGCCTCATATGTGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZIK1
PubMed Links
Gene Details
- Gene
- ZIK1
- Gene Name
- zinc finger protein interacting with K protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001010879.3 |
783 |
Silent Mutation |
AGA,AGG |
R147R |
NP_001010879.2 |
NM_001321145.1 |
783 |
Silent Mutation |
AGA,AGG |
R134R |
NP_001308074.1 |
NM_001321146.1 |
783 |
Silent Mutation |
AGA,AGG |
R92R |
NP_001308075.1 |
NM_001321147.1 |
783 |
Silent Mutation |
AGA,AGG |
R44R |
NP_001308076.1 |
XM_011526761.1 |
783 |
Silent Mutation |
AGA,AGG |
R133R |
XP_011525063.1 |
XM_011526762.1 |
783 |
Silent Mutation |
AGA,AGG |
R128R |
XP_011525064.1 |
XM_011526763.1 |
783 |
Silent Mutation |
AGA,AGG |
R120R |
XP_011525065.1 |
XM_011526764.1 |
783 |
Silent Mutation |
AGA,AGG |
R114R |
XP_011525066.1 |
XM_011526765.2 |
783 |
Silent Mutation |
AGA,AGG |
R111R |
XP_011525067.1 |
XM_011526766.2 |
783 |
Silent Mutation |
AGA,AGG |
R92R |
XP_011525068.1 |
XM_011526767.2 |
783 |
Silent Mutation |
AGA,AGG |
R44R |
XP_011525069.1 |
- Gene
- ZNF530
- Gene Name
- zinc finger protein 530
There are no transcripts associated with this gene.
View Full Product Details