Product Details

SNP ID
rs146194308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48873462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAATCCCCCATACCCCTTGGGGC[A/T]GACGCCCTGAAGAGGAGGCTGAAGA
Phenotype
MIM: 607769 MIM: 611048 MIM: 602309
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PLEKHA4 PubMed Links

Gene Details

Gene
PLEKHA4
Gene Name
pleckstrin homology domain containing A4
There are no transcripts associated with this gene.

Gene
PPP1R15A
Gene Name
protein phosphatase 1 regulatory subunit 15A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014330.3 498 Nonsense Mutation AGA,TGA R77* NP_055145.3
Gene
TULP2
Gene Name
tubby like protein 2
There are no transcripts associated with this gene.

View Full Product Details