Product Details

SNP ID
rs146269373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38628411 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCCAGCATCTCCATTCTCTGAC[C/T]GCTTCCTCCTAGCTTCTCAGCTCAC
Phenotype
MIM: 609596
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF3K PubMed Links

Gene Details

Gene
EIF3K
Gene Name
eukaryotic translation initiation factor 3 subunit K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300992.1 Intron NP_001287921.1
NM_001308393.1 Intron NP_001295322.1
NM_013234.3 Intron NP_037366.1

View Full Product Details