Product Details

SNP ID
rs146377382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14827462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGTTGCGGGTGGCAGCAGAACT[A/G]AGGTACACCCCTAGGATTGCACCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR7A5 PubMed Links

Gene Details

Gene
OR7A5
Gene Name
olfactory receptor family 7 subfamily A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017506.1 917 Silent Mutation CTC,CTT L260L NP_059976.1
XM_011527912.2 917 Silent Mutation CTC,CTT L260L XP_011526214.1
XM_011527913.2 917 Silent Mutation CTC,CTT L260L XP_011526215.1
XM_017026583.1 917 Silent Mutation CTC,CTT L304L XP_016882072.1
Gene
OR7C1
Gene Name
olfactory receptor family 7 subfamily C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198944.1 917 Intron NP_945182.1
XM_005259853.3 917 Intron XP_005259910.1
XM_011527915.2 917 Intron XP_011526217.1

View Full Product Details