Product Details

SNP ID
rs146780348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10274923 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGTTGCTCCTGCCTGGGAACAAC[C/T]GGAAGGTGTATGAACTGAGCAATGT
Phenotype
MIM: 147840
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ICAM1 PubMed Links

Gene Details

Gene
ICAM1
Gene Name
intercellular adhesion molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000201.2 545 Missense Mutation CGG,TGG R76W NP_000192.2

View Full Product Details