Product Details

SNP ID
rs147053389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15949675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCATCATCCTCTCCTATGTCTTCA[C/T]TGTGGCTGCCATCTTGAGGATTCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10H4 PubMed Links

Gene Details

Gene
OR10H4
Gene Name
olfactory receptor family 10 subfamily H member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004465.1 668 Missense Mutation ACT,ATT T223I NP_001004465.1

View Full Product Details