Product Details

SNP ID
rs147841193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12155420 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTTCACTTAAACATTCCTTCCTA[A/C]CCCTTTCAAATTTTAGACACAACTT
Phenotype
MIM: 604078
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF136 PubMed Links

Gene Details

Gene
ZNF136
Gene Name
zinc finger protein 136
There are no transcripts associated with this gene.

Gene
ZNF625
Gene Name
zinc finger protein 625
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145233.3 Intron NP_660276.2
Gene
ZNF625-ZNF20
Gene Name
ZNF625-ZNF20 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details