Product Details
- SNP ID
-
rs147847262
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:45872362 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCCCTGGCACACGCCAAGCCACC[A/G]TATTCCTATATCTCACTCATCACCA
- Phenotype
-
MIM: 602295
MIM: 602388
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FOXA3
PubMed Links
Gene Details
- Gene
- FOXA3
- Gene Name
- forkhead box A3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004497.2 |
554 |
Silent Mutation |
CCA,CCG |
P119P |
NP_004488.2 |
- Gene
- SYMPK
- Gene Name
- symplekin
There are no transcripts associated with this gene.
View Full Product Details