Product Details

SNP ID
rs148040275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42388981 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCTGTGTGCCAGTCACTGAGCTG[C/G]GTCTGGTAGCAGCTGGTGGTGGCGC
Phenotype
MIM: 611764
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CNFN PubMed Links

Gene Details

Gene
CNFN
Gene Name
cornifelin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032488.3 112 Silent Mutation ACC,ACG T19T NP_115877.2
XM_005259332.2 112 Silent Mutation ACC,ACG T32T XP_005259389.1
XM_011527396.1 112 Silent Mutation ACC,ACG T32T XP_011525698.1
XM_011527397.1 112 Silent Mutation ACC,ACG T32T XP_011525699.1
Gene
LIPE-AS1
Gene Name
LIPE antisense RNA 1
There are no transcripts associated with this gene.

Gene
LOC101930071
Gene Name
uncharacterized LOC101930071
There are no transcripts associated with this gene.

View Full Product Details