Product Details

SNP ID
rs148528739
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49583650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTCACACCAGGGAACCTGGAAC[A/G]GGAGTGTCTGGAAGAGAGGTGTTCC
Phenotype
MIM: 616759 MIM: 616633 MIM: 604429
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NOSIP PubMed Links

Gene Details

Gene
NOSIP
Gene Name
nitric oxide synthase interacting protein
There are no transcripts associated with this gene.

Gene
PRR12
Gene Name
proline rich 12
There are no transcripts associated with this gene.

Gene
PRRG2
Gene Name
proline rich and Gla domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000951.2 526 Missense Mutation CAG,CGG Q65R NP_000942.1
NM_001316335.1 526 Missense Mutation CAG,CGG Q42R NP_001303264.1
XM_006723286.2 526 Missense Mutation CAG,CGG Q65R XP_006723349.1
XM_011527124.1 526 Missense Mutation CAG,CGG Q42R XP_011525426.1

View Full Product Details