Product Details

SNP ID
rs148554460
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18784264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTGTCCTTCCAACCCACGTTTC[A/G]CGGGTCCTTCCACAGCAGCCGCACC
Phenotype
MIM: 600310 MIM: 607536
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COMP PubMed Links

Gene Details

Gene
COMP
Gene Name
cartilage oligomeric matrix protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000095.2 2050 Nonsense Mutation CGA,TGA R672* NP_000086.2
Gene
CRTC1
Gene Name
CREB regulated transcription coactivator 1
There are no transcripts associated with this gene.

View Full Product Details