Product Details
- SNP ID
-
rs148945232
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:35800072 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCCTTCGGCATCCTGGCAGCAGC[C/T]GCCGCCACAGTTCTTGGCTGAGCAG
- Phenotype
-
MIM: 616377
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LINC01529
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3761097] are located under a probe and SNP(s) [rs1133745] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LINC01529
- Gene Name
- long intergenic non-protein coding RNA 1529
There are no transcripts associated with this gene.
- Gene
- PRODH2
- Gene Name
- proline dehydrogenase 2
View Full Product Details