Product Details

SNP ID
rs149242446
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9960817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGTAGGTGAAGTTCTGGCGAGCT[A/G]TGGCACTCAGCAGTTTCAGGAAGTT
Phenotype
MIM: 120216
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL5A3 PubMed Links

Gene Details

Gene
COL5A3
Gene Name
collagen type V alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015719.3 2712 Missense Mutation ACA,ATA T1642I NP_056534.2
XM_011528042.2 2712 Missense Mutation ACA,ATA T1641I XP_011526344.1
XM_017026849.1 2712 Missense Mutation ACA,ATA T863I XP_016882338.1

View Full Product Details