Product Details

SNP ID
rs149297353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:20004708 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCTAATATAGAACTTCTCAAAC[C/G]CCGAGCAGCAGAAATTGACCACATG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF682 PubMed Links

Gene Details

Gene
ZNF682
Gene Name
zinc finger protein 682
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077349.1 3021 Intron NP_001070817.1
NM_033196.2 3021 Intron NP_149973.1
XM_017027455.1 3021 UTR 3 XP_016882944.1
XM_017027456.1 3021 Intron XP_016882945.1

View Full Product Details