Product Details

SNP ID
rs149327139
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46685569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAAGAGCGAGCAGGCAAAAAGA[A/G]AGGAAGAGATGAAGGCCAGAAAGAG
Phenotype
MIM: 607074
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DACT3-AS1 PubMed Links

Gene Details

Gene
DACT3-AS1
Gene Name
DACT3 antisense RNA 1
There are no transcripts associated with this gene.

Gene
PRKD2
Gene Name
protein kinase D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079880.1 Intron NP_001073349.1
NM_001079881.1 Intron NP_001073350.1
NM_001079882.1 Intron NP_001073351.1
NM_016457.4 Intron NP_057541.2
XM_005258716.2 Intron XP_005258773.2

View Full Product Details