Product Details
- SNP ID
-
rs149409914
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:39658546 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCCATTTGCGAGTGCACTTAGGCC[A/G]TCGTGTGGTCATGAACAGTCGTGAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LGALS16
PubMed Links
Gene Details
- Gene
- LGALS16
- Gene Name
- galectin 16
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001190441.2 |
247 |
Missense Mutation |
CAT,CGT |
H60R |
NP_001177370.2 |
View Full Product Details