Product Details

SNP ID
rs149457093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52613135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATGTTTCTCTCCAGTGTGGATC[A/G]CATGATGATTAGTGAGGCTTGAACG
Phenotype
MIM: 194558
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC107983997 PubMed Links

Gene Details

Gene
LOC107983997
Gene Name
zinc finger protein 816-like
There are no transcripts associated with this gene.

Gene
ZNF701
Gene Name
zinc finger protein 701
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172655.1 2011 Intron NP_001166126.1
NM_018260.2 2011 Intron NP_060730.2
XM_011527092.1 2011 Intron XP_011525394.1
Gene
ZNF83
Gene Name
zinc finger protein 83
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105549.1 2011 Intron NP_001099019.1
NM_001105550.1 2011 Intron NP_001099020.1
NM_001105551.1 2011 Intron NP_001099021.1
NM_001105552.1 2011 Intron NP_001099022.1
NM_001277945.1 2011 Missense Mutation GCG,GTG A477V NP_001264874.1
NM_001277946.1 2011 Missense Mutation GCG,GTG A477V NP_001264875.1
NM_001277947.1 2011 Missense Mutation GCG,GTG A477V NP_001264876.1
NM_001277948.1 2011 Missense Mutation GCG,GTG A477V NP_001264877.1
NM_001277949.1 2011 Missense Mutation GCG,GTG A477V NP_001264878.1
NM_001277951.1 2011 Intron NP_001264880.1
NM_001277952.1 2011 Intron NP_001264881.1
NM_018300.3 2011 Intron NP_060770.3
XM_017026951.1 2011 Missense Mutation GCG,GTG A477V XP_016882440.1

View Full Product Details