Product Details

SNP ID
rs149625707
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45008865 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTTGTCCTGTCTATGGAATGAC[A/G]GGAGGTCGGTTTCGATTCCTCCACG
Phenotype
MIM: 604758
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RELB PubMed Links

Gene Details

Gene
RELB
Gene Name
RELB proto-oncogene, NF-kB subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006509.3 Intron NP_006500.2
XM_005259127.3 Intron XP_005259184.1
XM_005259128.2 Intron XP_005259185.1

View Full Product Details