Product Details

SNP ID
rs149707549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58335048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTCAGTGGCTGCAGCCCGAGGC[A/G]CACTCCAAGGAGCAGATACTGGAGC
Phenotype
MIM: 165260
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6806 PubMed Links

Gene Details

Gene
MIR6806
Gene Name
microRNA 6806
There are no transcripts associated with this gene.

Gene
ZSCAN22
Gene Name
zinc finger and SCAN domain containing 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321116.1 393 Silent Mutation GCA,GCG A82A NP_001308045.1
NM_001321117.1 393 Silent Mutation GCA,GCG A82A NP_001308046.1
NM_181846.2 393 Silent Mutation GCA,GCG A82A NP_862829.1
XM_006723192.3 393 Silent Mutation GCA,GCG A82A XP_006723255.1
XM_011526917.2 393 Silent Mutation GCA,GCG A82A XP_011525219.1
XM_011526918.2 393 Intron XP_011525220.1
XM_017026746.1 393 Silent Mutation GCA,GCG A82A XP_016882235.1
XM_017026747.1 393 UTR 5 XP_016882236.1

View Full Product Details