Product Details

SNP ID
rs149895971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41958347 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAATGTAACAGGCGCCGAAAAA[C/G]ACAGCCAGAGCCACCAGCAACATAG
Phenotype
MIM: 182350 MIM: 604925
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATP1A3 PubMed Links

Gene Details

Gene
ATP1A3
Gene Name
ATPase Na+/K+ transporting subunit alpha 3
There are no transcripts associated with this gene.

Gene
RABAC1
Gene Name
Rab acceptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006423.2 390 Silent Mutation GTC,GTG V102V NP_006414.2

View Full Product Details