Product Details

SNP ID
rs149947768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44287384 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGCCCTTCCCACACTGCTGACAC[A/G]TATAGGGTTTCTCTCCTGTGTGGAC
Phenotype
MIM: 604749
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF235 PubMed Links

Gene Details

Gene
ZNF235
Gene Name
zinc finger protein 235
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004234.4 6210 Missense Mutation ACG,ATG T684M NP_004225.3
XM_005259412.4 6210 Missense Mutation ACG,ATG T680M XP_005259469.1

View Full Product Details