Product Details

SNP ID
rs150091315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43511563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCAGGGCTGGCCCTGGTCTCCA[A/G]CGCCTGGCAGGGGTGGAAGAGTACA
Phenotype
MIM: 608451
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ETHE1 PubMed Links

Gene Details

Gene
ETHE1
Gene Name
ETHE1, persulfide dioxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320867.1 164 Silent Mutation CTG,TTG L116L NP_001307796.1
NM_001320868.1 164 Silent Mutation CTG,TTG L4L NP_001307797.1
NM_001320869.1 164 Silent Mutation CTG,TTG L29L NP_001307798.1
NM_014297.4 164 Silent Mutation CTG,TTG L127L NP_055112.2
XM_005258687.3 164 Silent Mutation CTG,TTG L100L XP_005258744.1
Gene
PHLDB3
Gene Name
pleckstrin homology like domain family B member 3
There are no transcripts associated with this gene.

View Full Product Details