Product Details

SNP ID
rs150212158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49930910 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGGCCCTGCTCCCAGCTAGTGG[A/G]CTGGGATGGCTCGTAGACTATGGGA
Phenotype
MIM: 606398 MIM: 609742 MIM: 605815
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATF5 PubMed Links

Gene Details

Gene
ATF5
Gene Name
activating transcription factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193646.1 437 Silent Mutation GGA,GGG G20G NP_001180575.1
NM_001290746.1 437 Silent Mutation GGA,GGG G20G NP_001277675.1
NM_012068.5 437 Silent Mutation GGA,GGG G20G NP_036200.2
XM_011526629.2 437 Silent Mutation GGA,GGG G20G XP_011524931.1
Gene
IL4I1
Gene Name
interleukin 4 induced 1
There are no transcripts associated with this gene.

Gene
MIR4751
Gene Name
microRNA 4751
There are no transcripts associated with this gene.

Gene
NUP62
Gene Name
nucleoporin 62
There are no transcripts associated with this gene.

View Full Product Details