Product Details

SNP ID
rs150226322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50661598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCCGAGGGCACCCCCTTCCCCTG[C/T]GGCCTGGGGGACAGTCCCCATCCAA
Phenotype
MIM: 604999 MIM: 600327
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C19orf81 PubMed Links

Gene Details

Gene
C19orf81
Gene Name
chromosome 19 open reading frame 81
There are no transcripts associated with this gene.

Gene
SHANK1
Gene Name
SH3 and multiple ankyrin repeat domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016148.3 6872 UTR 3 NP_057232.2
XM_006723233.3 6872 UTR 3 XP_006723296.1
XM_011527013.2 6872 UTR 3 XP_011525315.1
XM_011527014.2 6872 UTR 3 XP_011525316.1
Gene
SYT3
Gene Name
synaptotagmin 3
There are no transcripts associated with this gene.

View Full Product Details