Product Details

SNP ID
rs150845309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40667236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGGGCATTGCCAGCCTAAGTCC[A/G]GCAGTGAAATGGTTCCCTTAGCCAG
Phenotype
MIM: 604018
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUMBL PubMed Links

Gene Details

Gene
NUMBL
Gene Name
NUMB like, endocytic adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289979.1 2152 UTR 3 NP_001276908.1
NM_001289980.1 2152 UTR 3 NP_001276909.1
NM_004756.4 2152 UTR 3 NP_004747.1

View Full Product Details