Product Details

SNP ID
rs151053559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15742259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCTTCACCCCCTTCCTCAGCCC[A/G]ATCATTTTCAGTCTAAGGAACAAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10H3 PubMed Links

Gene Details

Gene
OR10H3
Gene Name
olfactory receptor family 10 subfamily H member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013938.1 867 Silent Mutation CCA,CCG P289P NP_039226.1

View Full Product Details