Product Details

SNP ID
rs111493048
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:40988640 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGAAAGCATGCCCTTTATTGAGG[C/T]CTTCCGTCAGTTCCAATTCAAGGTC
Phenotype
MIM: 123860
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTPS1 PubMed Links

Gene Details

Gene
CTPS1
Gene Name
CTP synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301237.1 1029 Missense Mutation GCC,GTC A6V NP_001288166.1
NM_001905.3 1029 Missense Mutation GCC,GTC A162V NP_001896.2

View Full Product Details