Product Details

SNP ID
rs111543981
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:233905711 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTCCATCGAGGAGCGGATCCTGC[C/G]CATCACTGGCTACTATGGCTACCAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC35F3 PubMed Links

Gene Details

Gene
SLC35F3
Gene Name
solute carrier family 35 member F3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300845.1 602 Intron NP_001287774.1
NM_173508.3 602 Missense Mutation CCC,CGC P79R NP_775779.1

View Full Product Details