Product Details

SNP ID
rs111568119
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:27357124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGATCTGCTTCACCTGCAGGGG[A/G]AGGGAGGCGCCGCTGAGACAGCTGA
Phenotype
MIM: 604468 MIM: 608042
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAP3K6 PubMed Links

Gene Details

Gene
MAP3K6
Gene Name
mitogen-activated protein kinase kinase kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297609.1 Intron NP_001284538.1
NM_004672.4 Intron NP_004663.3
XM_017002771.1 Intron XP_016858260.1
XM_017002772.1 Intron XP_016858261.1
Gene
SYTL1
Gene Name
synaptotagmin like 1
There are no transcripts associated with this gene.

View Full Product Details