Product Details

SNP ID
rs111742567
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:153548800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCCACCGTGGGGCTCCTTAAGC[A/G]CTCTCAGATTATCTGCCACCTCTTC
Phenotype
MIM: 176992 MIM: 114210 MIM: 176991
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928034 PubMed Links

Gene Details

Gene
LOC101928034
Gene Name
uncharacterized LOC101928034
There are no transcripts associated with this gene.

Gene
S100A3
Gene Name
S100 calcium binding protein A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002960.1 Intron NP_002951.1
Gene
S100A4
Gene Name
S100 calcium binding protein A4
There are no transcripts associated with this gene.

Gene
S100A5
Gene Name
S100 calcium binding protein A5
There are no transcripts associated with this gene.

View Full Product Details