Product Details
- SNP ID
-
rs111803813
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:31364132 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGAGGGCAAGGCACACATCAAAAG[A/G]CAGCAAGGCAGCAAAGAAGAAGAAA
- Phenotype
-
MIM: 134651
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FABP3
PubMed Links
Gene Details
- Gene
- FABP3
- Gene Name
- fatty acid binding protein 3
- Gene
- ZCCHC17
- Gene Name
- zinc finger CCHC-type containing 17
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001282566.1 |
958 |
Missense Mutation |
GAC,GGC |
D244G |
NP_001269495.1 |
NM_001282567.1 |
958 |
Missense Mutation |
GAC,GGC |
D224G |
NP_001269496.1 |
NM_001282568.1 |
958 |
Missense Mutation |
GAC,GGC |
D222G |
NP_001269497.1 |
NM_001282569.1 |
958 |
Missense Mutation |
GAC,GGC |
D214G |
NP_001269498.1 |
NM_001282570.1 |
958 |
Missense Mutation |
GAC,GGC |
D198G |
NP_001269499.1 |
NM_001282571.1 |
958 |
Missense Mutation |
GAC,GGC |
D160G |
NP_001269500.1 |
NM_001282572.1 |
958 |
Missense Mutation |
ACA,GCA |
T140A |
NP_001269501.1 |
NM_001282573.1 |
958 |
Missense Mutation |
ACA,GCA |
T132A |
NP_001269502.1 |
NM_001282574.1 |
958 |
UTR 3 |
|
|
NP_001269503.1 |
NM_016505.3 |
958 |
Missense Mutation |
GAC,GGC |
D222G |
NP_057589.2 |
XM_006710681.3 |
958 |
Missense Mutation |
GAC,GGC |
D214G |
XP_006710744.1 |
XM_011541567.2 |
958 |
Missense Mutation |
GAC,GGC |
D222G |
XP_011539869.1 |
View Full Product Details