Product Details

SNP ID
rs111907007
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:53949640 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGAGGAACAGCTCAGAATAAAT[A/G]AGGTGTCTCCTTAGCAGCAGGGGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSPB11 PubMed Links

Gene Details

Gene
HSPB11
Gene Name
heat shock protein family B (small) member 11
There are no transcripts associated with this gene.

Gene
LRRC42
Gene Name
leucine rich repeat containing 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256409.1 Intron NP_001243338.1
NM_052940.4 Intron NP_443172.1
XM_006710327.3 Intron XP_006710390.1
XM_006710328.3 Intron XP_006710391.1

View Full Product Details