Product Details

SNP ID
rs112176770
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:44806917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGGGATCCTCGGCTAGTCCTTG[A/C]CGACTGAGCTCCTTCTCTGGGCACT
Phenotype
MIM: 602913 MIM: 611713
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BTBD19 PubMed Links

Gene Details

Gene
BTBD19
Gene Name
BTB domain containing 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136537.1 369 Intron NP_001130009.1
XM_006710386.3 369 Intron XP_006710449.1
XM_011540803.2 369 Intron XP_011539105.1
XM_011540805.2 369 Intron XP_011539107.1
XM_017000447.1 369 Intron XP_016855936.1
XM_017000448.1 369 Intron XP_016855937.1
XM_017000449.1 369 Intron XP_016855938.1
Gene
PLK3
Gene Name
polo like kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004073.3 369 Intron NP_004064.2
Gene
TCTEX1D4
Gene Name
Tctex1 domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013632.3 369 UTR 5 NP_001013654.1
XM_006710614.3 369 Intron XP_006710677.1
XM_011541375.2 369 Intron XP_011539677.1
XM_011541376.2 369 Intron XP_011539678.1
XM_017001187.1 369 Intron XP_016856676.1

View Full Product Details