Product Details

SNP ID
rs112412366
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:161230102 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGTTCCAAAGGTCCTCCAGGGG[G/T]CCTCGGTCTGACACTGTCTTCTCTC
Phenotype
MIM: 107670 MIM: 603881
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
APOA2 PubMed Links

Gene Details

Gene
APOA2
Gene Name
apolipoprotein A2
There are no transcripts associated with this gene.

Gene
MIR5187
Gene Name
microRNA 5187
There are no transcripts associated with this gene.

Gene
NR1I3
Gene Name
nuclear receptor subfamily 1 group I member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077469.2 2267 Intron NP_001070937.1
NM_001077470.2 2267 Intron NP_001070938.1
NM_001077471.2 2267 Intron NP_001070939.1
NM_001077472.2 2267 Intron NP_001070940.1
NM_001077473.2 2267 Intron NP_001070941.1
NM_001077474.2 2267 Intron NP_001070942.1
NM_001077475.2 2267 Intron NP_001070943.1
NM_001077476.2 2267 Intron NP_001070944.1
NM_001077477.2 2267 Intron NP_001070945.1
NM_001077478.2 2267 Intron NP_001070946.1
NM_001077479.2 2267 Intron NP_001070947.1
NM_001077480.2 2267 Intron NP_001070948.1
NM_001077481.2 2267 Intron NP_001070949.1
NM_001077482.2 2267 Intron NP_001070950.1
NM_005122.4 2267 Intron NP_005113.1
XM_005245693.4 2267 Intron XP_005245750.1
XM_005245694.4 2267 Intron XP_005245751.1
XM_005245697.4 2267 Intron XP_005245754.1
XM_011510237.2 2267 Intron XP_011508539.1
Gene
TOMM40L
Gene Name
translocase of outer mitochondrial membrane 40 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286373.1 2267 UTR 3 NP_001273302.1
NM_001286374.1 2267 UTR 3 NP_001273303.1
NM_032174.5 2267 UTR 3 NP_115550.2
XM_006711572.2 2267 UTR 3 XP_006711635.1
XM_011510057.2 2267 UTR 3 XP_011508359.1
XM_017002481.1 2267 UTR 3 XP_016857970.1
XM_017002482.1 2267 UTR 3 XP_016857971.1

View Full Product Details