Product Details

SNP ID
rs112687190
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:23193355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCTCGAGCAGCAGAAATCCTCT[A/T]GCGTTCCAGGGCACTGTCAGCAAGC
Phenotype
MIM: 182133
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HTR1D PubMed Links

Gene Details

Gene
HTR1D
Gene Name
5-hydroxytryptamine receptor 1D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000864.4 1375 Nonsense Mutation AAG,TAG K289* NP_000855.1

View Full Product Details