Product Details

SNP ID
rs113210734
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:27322242 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTCTGCTCTTGTTGCCGCCGCC[A/G]CCACCCCCGCCCAGGATGGCGGAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTG1P20 PubMed Links

Gene Details

Gene
ACTG1P20
Gene Name
actin gamma 1 pseudogene 20
There are no transcripts associated with this gene.

Gene
TMEM222
Gene Name
transmembrane protein 222
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032125.2 98 Silent Mutation CCA,CCG P15P NP_115501.2

View Full Product Details