Product Details

SNP ID
rs113247024
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:6461439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGCAAGTGGGCGCCGTGTCACG[C/G]GCCGCGCTGCAGGCGGCTGCGCAAG
Phenotype
MIM: 606351 MIM: 611101 MIM: 603366
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ESPN PubMed Links

Gene Details

Gene
ESPN
Gene Name
espin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031475.2 1226 Intron NP_113663.2
XM_005263501.2 1226 Intron XP_005263558.1
XM_011542231.1 1226 Intron XP_011540533.1
XM_011542232.1 1226 Intron XP_011540534.1
XM_011542233.2 1226 Intron XP_011540535.1
XM_011542236.2 1226 Intron XP_011540538.1
XM_011542237.1 1226 Intron XP_011540539.1
XM_011542238.2 1226 Intron XP_011540540.1
XM_017002433.1 1226 Intron XP_016857922.1
XM_017002434.1 1226 Intron XP_016857923.1
Gene
PLEKHG5
Gene Name
pleckstrin homology and RhoGEF domain containing G5
There are no transcripts associated with this gene.

Gene
TNFRSF25
Gene Name
TNF receptor superfamily member 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039664.1 1226 Intron NP_001034753.1
NM_003790.2 1226 Missense Mutation CCG,GCG P417A NP_003781.1
NM_148965.1 1226 Missense Mutation CCG,GCG P426A NP_683866.1
NM_148966.1 1226 Missense Mutation CCG,GCG P380A NP_683867.1
NM_148967.1 1226 Missense Mutation CCG,GCG P372A NP_683868.1
NM_148970.1 1226 Missense Mutation CCG,GCG P234A NP_683871.1

View Full Product Details