Product Details

SNP ID
rs113655390
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:32863065 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTTTCAGTTGTCCAAGCTAGCA[C/T]TTCTGAAACTCCAAGAGATACTTCC
Phenotype
MIM: 611906 MIM: 611889
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FNDC5 PubMed Links

Gene Details

Gene
FNDC5
Gene Name
fibronectin type III domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171940.1 1935 UTR 3 NP_001165411.2
NM_001171941.2 1935 UTR 3 NP_001165412.1
NM_153756.2 1935 UTR 3 NP_715637.2
Gene
S100PBP
Gene Name
S100P binding protein
There are no transcripts associated with this gene.

View Full Product Details