Product Details

SNP ID
rs113707713
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:109250541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCCTGCAAGCTGGCACAGGCCCC[A/C]GGGCTCAGGGCAGGGGAAAGGTCAC
Phenotype
MIM: 604265
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CELSR2 PubMed Links

Gene Details

Gene
CELSR2
Gene Name
cadherin EGF LAG seven-pass G-type receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001408.2 523 Silent Mutation CCA,CCC P154P NP_001399.1
XM_005270580.4 523 Silent Mutation CCA,CCC P154P XP_005270637.1

View Full Product Details