Product Details
- SNP ID
-
rs113941793
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
15
- Location
-
Chr.1:207887761 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTCACAATTCGGTATCAGCCACCA[C/T]GTGTTGTCTTGCTGAATGGCCGTTT
- Phenotype
-
MIM: 142230
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD34
PubMed Links
Gene Details
- Gene
- CD34
- Gene Name
- CD34 molecule
View Full Product Details