Product Details

SNP ID
rs114240437
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:54805181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGCAGGGCCTCTGGTACAACCTA[C/T]GCCTCTCTGGCTTCAATCCTGTCAC
Phenotype
MIM: 616446
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LEXM PubMed Links

Gene Details

Gene
LEXM
Gene Name
lymphocyte expansion molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110533.1 Intron NP_001104003.1
NM_152607.2 Intron NP_689820.2
XM_017000480.1 Intron XP_016855969.1
XM_017000481.1 Intron XP_016855970.1
Gene
TTC22
Gene Name
tetratricopeptide repeat domain 22
There are no transcripts associated with this gene.

View Full Product Details