Product Details

SNP ID
rs114257826
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:44227200 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGTCTGCTGGAGTCTCCACAGG[C/T]CTCCTGCCTGCAGAATGCTTAGCTT
Phenotype
MIM: 605077 MIM: 609917
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DMAP1 PubMed Links

Gene Details

Gene
DMAP1
Gene Name
DNA methyltransferase 1 associated protein 1
There are no transcripts associated with this gene.

Gene
ERI3
Gene Name
ERI1 exoribonuclease family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301698.1 Intron NP_001288627.1
NM_001301699.1 Intron NP_001288628.1
NM_001301700.1 Intron NP_001288629.1
NM_001301701.1 Intron NP_001288630.1
NM_024066.2 Intron NP_076971.1
XM_005271186.2 Intron XP_005271243.1
XM_006710891.3 Intron XP_006710954.1
XM_017002301.1 Intron XP_016857790.1
XM_017002302.1 Intron XP_016857791.1
XM_017002303.1 Intron XP_016857792.1
XM_017002304.1 Intron XP_016857793.1
XM_017002305.1 Intron XP_016857794.1
Gene
ERI3-IT1
Gene Name
ERI3 intronic transcript 1
There are no transcripts associated with this gene.

View Full Product Details