Product Details

SNP ID
rs114544019
Assay Type
Functionally Tested
NCBI dbSNP Submissions
14
Location
Chr.1:175070161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCTTGCTGTGAGAAAGTGATTC[A/G]TTCTCTGGGGGTAAATGAACATCTT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TNN PubMed Links

Gene Details

Gene
TNN
Gene Name
tenascin N
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022093.1 Intron NP_071376.1
XM_017002048.1 Intron XP_016857537.1
XM_017002049.1 Intron XP_016857538.1

View Full Product Details