Product Details

SNP ID
rs114629222
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:110405998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGAGCCTAGCAGTCTCAGCATAT[A/G]GCATACATTCAATAAATGGTTAACA
Phenotype
MIM: 608521
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAMTOR5 PubMed Links

Gene Details

Gene
LAMTOR5
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006402.2 Intron NP_006393.2
Gene
LAMTOR5-AS1
Gene Name
LAMTOR5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details