Product Details

SNP ID
rs114674626
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:226152531 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTCCTCGGCCCACTGTAATCAC[A/G]GAATCTGCATCCTGCTGAATCTTCT
Phenotype
MIM: 606809
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACBD3 PubMed Links

Gene Details

Gene
ACBD3
Gene Name
acyl-CoA binding domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022735.3 1226 Silent Mutation TCC,TCT S393S NP_073572.2

View Full Product Details