Product Details

SNP ID
rs114747108
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:180634707 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGTTTCTAGCCTAGGGTGCTTTC[C/T]AGTATCGTGAATCAATGTGAACTGT
Phenotype
MIM: 605237
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
XPR1 PubMed Links

Gene Details

Gene
XPR1
Gene Name
xenotropic and polytropic retrovirus receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135669.1 Intron NP_001129141.1
NM_004736.3 Intron NP_004727.2

View Full Product Details