Product Details
- SNP ID
-
rs114790584
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
8
- Location
-
Chr.1:157092294 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCCCAGACAAGACTCAGGGAACA[C/T]AACCACCCCTGTCCTCTCCCCAGGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ETV3L
PubMed Links
Gene Details
- Gene
- ETV3L
- Gene Name
- ETS variant 3 like
View Full Product Details