Product Details

SNP ID
rs114855019
Assay Type
Functionally Tested
NCBI dbSNP Submissions
13
Location
Chr.1:184398896 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTGCTGACCAAAACATGGTTAT[A/G]TGGCACATGGCTGTATGTATTTCTA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C1orf21 PubMed Links

Gene Details

Gene
C1orf21
Gene Name
chromosome 1 open reading frame 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030806.3 Intron NP_110433.1

View Full Product Details