Product Details

SNP ID
rs114941235
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:113881987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGATTCCACAGTGTCCTGGAGT[A/G]TAGCTCCTGTCTGAGGAAAGACAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP4B1-AS1 PubMed Links

Gene Details

Gene
AP4B1-AS1
Gene Name
AP4B1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
BCL2L15
Gene Name
BCL2 like 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010922.2 432 Missense Mutation ACA,ATA T87I NP_001010922.1

View Full Product Details